Zirconium standard solution, 1000 mg/L Zr in 5% hydrochloric acid (from ZrOCl₂), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13875.AD
13875.AE
13875.AP
13875.ADEA
53
EUR
13875.AD
13875.AE
13875.AP
Zirconium standard solution, 1000 mg/L Zr in 5% hydrochloric acid (from ZrOCl₂), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Zirkonium standaarden
Molecular Formula: ZrCl₂O in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011303
Certificaten: Traceable to NIST