Calcium standard solution, 1000 mg/L Ca in 5% nitric acid (from CaCO₃), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88057.AE 88057.AP
88057.AEEA 40 EUR
88057.AE 88057.AP
Calcium standard solution, 1000 mg/L Ca in 5% nitric acid (from CaCO₃), standard for AAS, Specpure®
Standaarden Kationische standaarden Calcium standaarden
Molecular Formula: CaCO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085314
Certificaten: Traceable to NIST