Erbium standard solution, 10000 mg/L Er in 5% nitric acid (from Er₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14409.AD
14409.AE
14409.AP
14409.ADEA
102
EUR
14409.AD
14409.AE
14409.AP
Erbium standard solution, 10000 mg/L Er in 5% nitric acid (from Er₂O₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Erbium standaarden
Molecular Formula: Er₂O₃ in 5% HNO₃ Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010987
Certificaten: Traceable to NIST