Erbium standard solution, 1000 mg/L Er in 5% nitric acid (from Er₂O₃), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88063.AE 88063.AP
88063.AEEA 44.4 EUR
88063.AE 88063.AP
Erbium standard solution, 1000 mg/L Er in 5% nitric acid (from Er₂O₃), standard for AAS, Specpure®
Standaarden Kationische standaarden Erbium standaarden
Molecular Formula: Er₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010987
Certificaten: Traceable to NIST