Europium standard solution, 10000 mg/L Eu in 5% nitric acid (from Eu?O?), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35761.AD 35761.AE 35761.AP
35761.ADEA 97.1 EUR
35761.AD 35761.AE 35761.AP
Europium standard solution, 10000 mg/L Eu in 5% nitric acid (from Eu?O?), standard for ICP, Specpure®
Standaarden Kationische standaarden Europium standaarden
Molecular Formula: Eu₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010992
Certificaten: Traceable to NIST