Iridium standard solution, 10000 mg/L Ir in 20% hydrochloric acid (from IrCl₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35752.AD
35752.AE
35752.AP
35752.ADEA
398
EUR
35752.AD
35752.AE
35752.AP
Iridium standard solution, 10000 mg/L Ir in 20% hydrochloric acid (from IrCl₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Iridium standaarden
Molecular Formula: IrCl₃ in 20% HCl
Physical Form: Liquid
Odour: Characteristic
UN No.: 1789
MDL No.: MFCD00011062
Certificaten: Traceable to NIST