Iridium standard solution, 1000 mg/L Ir in 20% hydrochloric acid (from IrCl₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35751.AD 35751.AE 35751.AP
35751.ADEA 109 EUR
35751.AD 35751.AE 35751.AP
Iridium standard solution, 1000 mg/L Ir in 20% hydrochloric acid (from IrCl₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Iridium standaarden
Molecular Formula: IrCl₃ in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011062
Certificaten: Traceable to NIST