Iridium standard solution, 1000 mg/L Ir in 20% hydrochloric acid (from IrCl₃), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88072.AE 88072.AP
88072.AEEA 121 EUR
88072.AE 88072.AP
Iridium standard solution, 1000 mg/L Ir in 20% hydrochloric acid (from IrCl₃), standard for AAS, Specpure®
Standaarden Kationische standaarden Iridium standaarden
Molecular Formula: IrCl₃ in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011062
Certificaten: Traceable to NIST