Potassium standard solution, 10000 mg/L K in 5% nitric acid (from KNO₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14379.AD
14379.AE
14379.AP
14379.ADEA
95.9
EUR
14379.AD
14379.AE
14379.AP
Potassium standard solution, 10000 mg/L K in 5% nitric acid (from KNO₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Kalium standaarden
Molecular Formula: KNO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00133776
Certificaten: Traceable to NIST