Potassium standard solution, 1000 mg/L K in 5% nitric acid (from KNO₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13866.AD 13866.AE 13866.AP
13866.ADEA 51.5 EUR
13866.AD 13866.AE 13866.AP
Potassium standard solution, 1000 mg/L K in 5% nitric acid (from KNO₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Kalium standaarden
Molecular Formula: KNO₃ in 5% HNO₃ Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00133776
Certificaten: Traceable to NIST