Potassium standard solution, 1000 mg/L K in 5% nitric acid (from KNO₃), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88087.AE
88087.AP
88087.AEEA
38.9
EUR
88087.AE
88087.AP
Potassium standard solution, 1000 mg/L K in 5% nitric acid (from KNO₃), standard for AAS, Specpure®
Standaarden
Kationische standaarden
Kalium standaarden
Molecular Formula: KNO₃ 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00133776
Certificaten: Traceable to NIST