Copper standard solution, 10000 mg/L Cu in 5% nitric acid (from Cu), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14372.AD
14372.AE
14372.AP
14372.ADEA
90.7
EUR
14372.AD
14372.AE
14372.AP
Copper standard solution, 10000 mg/L Cu in 5% nitric acid (from Cu), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Koper standaarden
Molecular Formula: Cu in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010965
Certificaten: Traceable to NIST