Copper standard solution, 1000 mg/L Cu in 5% nitric acid (from Cu), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88061.AE
88061.AP
88061.AEEA
40.9
EUR
88061.AE
88061.AP
Copper standard solution, 1000 mg/L Cu in 5% nitric acid (from Cu), standard for AAS, Specpure®
Standaarden
Kationische standaarden
Koper standaarden
Molecular Formula: Cu in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010965
Certificaten: Traceable to NIST