Lanthanum standard solution, 1000 mg/L La in 5% nitric acid (from La₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13870.AD
13870.AE
13870.AP
13870.ADEA
51.1
EUR
13870.AD
13870.AE
13870.AP
Lanthanum standard solution, 1000 mg/L La in 5% nitric acid (from La₂O₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Lanthanium standaarden
Molecular Formula: La₂O₃ in 5% HNO₃ Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011066
Certificaten: Traceable to NIST