Lutetium standard solution, 1000 mg/L Lu in 5% nitric acid (from Lu₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35765.AD
35765.AE
35765.AP
35765.ADEA
175
EUR
35765.AD
35765.AE
35765.AP
Lutetium standard solution, 1000 mg/L Lu in 5% nitric acid (from Lu₂O₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Lutetium standaarden
Molecular Formula: Lu₂O3 in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011098
Certificaten: Traceable to NIST