Lutetium standard solution, 1000 mg/L Lu in 5% nitric acid (from Lu₂O₃), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
89886.AE
89886.AP
89886.AEEA
65.6
EUR
89886.AE
89886.AP
Lutetium standard solution, 1000 mg/L Lu in 5% nitric acid (from Lu₂O₃), standard for AAS, Specpure®
Standaarden
Kationische standaarden
Lutetium standaarden
Molecular Formula: Lu₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011098
Certificaten: Traceable to NIST