Neodymium standard solution, 10000 mg/L Nd in 5% nitric acid (from Nd₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14431.AE
14431.AD
14431.AP
14431.AEEA
167
EUR
14431.AE
14431.AD
14431.AP
Neodymium standard solution, 10000 mg/L Nd in 5% nitric acid (from Nd₂O₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Neodymium standaarden
Molecular Formula: Nd₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011130
Certificaten: Traceable to NIST