Neodymium standard solution, 1000 mg/L Nd in 5% nitric acid (from Nd₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13882.AD
13882.AE
13882.AP
13882.ADEA
72.5
EUR
13882.AD
13882.AE
13882.AP
Neodymium standard solution, 1000 mg/L Nd in 5% nitric acid (from Nd₂O₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Neodymium standaarden
Molecular Formula: Nd₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011130
Certificaten: Traceable to NIST