Rubidium standard solution, 10000 mg/L Rb in 5% nitric acid (from Rb₂CO₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14380.AE
14380.AD
14380.AP
14380.AEEA
124
EUR
14380.AE
14380.AD
14380.AP
Rubidium standard solution, 10000 mg/L Rb in 5% nitric acid (from Rb₂CO₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Rubidium standaarden
Molecular Formula: Rb₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134055
Certificaten: Traceable to NIST