Rubidium standard solution, 1000 mg/L Rb in 5% nitric acid (from Rb₂CO₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13872.ADEA 52 EUR
13872.AD 13872.AE
Rubidium standard solution, 1000 mg/L Rb in 5% nitric acid (from Rb₂CO₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Rubidium standaarden
Molecular Formula: Rb₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134055
Certificaten: Traceable to NIST