Scandium standard solution, 10000 mg/L Sc in 5% nitric acid (from Sc₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35755.AD
35755.AE
35755.AP
35755.ADEA
265
EUR
35755.AD
35755.AE
35755.AP
Scandium standard solution, 10000 mg/L Sc in 5% nitric acid (from Sc₂O₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Scandium standaarden
Molecular Formula: Sc₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00016323
Certificaten: Traceable to NIST