Selenium standard solution, 10000 mg/L Se in 5% nitric acid (from Se), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14381.AD
14381.AE
14381.AP
14381.ADEA
85.7
EUR
14381.AD
14381.AE
14381.AP
Selenium standard solution, 10000 mg/L Se in 5% nitric acid (from Se), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Selenium standaarden
Molecular Formula: Se in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134090
Certificaten: Traceable to NIST