Selenium standard solution, 1000 mg/L Se in 5% nitric acid (from Se), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88094.AE 88094.AP
88094.AEEA 38.6 EUR
88094.AE 88094.AP
Selenium standard solution, 1000 mg/L Se in 5% nitric acid (from Se), standard for AAS, Specpure®
Standaarden Kationische standaarden Selenium standaarden
Molecular Formula: Se in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134090
Certificaten: Traceable to NIST