Thallium standard solution, 1000 mg/L Tl in 5% nitric acid (from Tl), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13851.AD 13851.AE 13851.AP
13851.ADEA 51.1 EUR
13851.AD 13851.AE 13851.AP
Thallium standard solution, 1000 mg/L Tl in 5% nitric acid (from Tl), standard for ICP, Specpure®
Standaarden Kationische standaarden Thallium standaarden
Molecular Formula: Tl in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134063
Certificaten: Traceable to NIST