Tin standard solution, 10000 mg/L Sn in 20% hydrochloric acid (from Sn), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14384.AD 14384.AE 14384.AP
14384.ADEA 102 EUR
14384.AD 14384.AE 14384.AP
Tin standard solution, 10000 mg/L Sn in 20% hydrochloric acid (from Sn), standard for ICP, Specpure®
Standaarden Kationische standaarden Tin standaarden
Molecular Formula: Sn in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00133862
Certificaten: Traceable to NIST