Chromium standard solution, 10000 mg/L Cr in 5% hydrochloric acid (from Cr), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14408.AD 14408.AE 14408.AP
14408.ADEA 88 EUR
14408.AD 14408.AE 14408.AP
Chromium standard solution, 10000 mg/L Cr in 5% hydrochloric acid (from Cr), standard for ICP, Specpure®
Standaarden Kationische standaarden Chroom standaarden
Molecular Formula: Cr in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00010944
Certificaten: Traceable to NIST