trans,trans-2,4-Hexadienal predominantly trans 95%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 95%
CAS No.: 142-83-6
Molecular Formula: C6H8O
Formula Weight: 96.13
Storage Temperature: 2°C to 8°C
Flash Point: 67°(152°F)
Refractive Index: 1.5386
Odour: Floral
Boiling Point: 170-173°
Beilstein Reference: 1698401
UN No.: 2922
MDL No.: MFCD00007004
Formule:
C₆H₈O Kookpunt: 170…173 °C Dichtheid: 0,871 Vlampunt: 67 °C (152 °F) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00007004 CAS nummer: 142-83-6 EINECS: 205-564-3 UN: 2922 ADR: 8,III |