Germanium standard solution, 1000 mg/L Ge in 5% nitric acid with hydrofluoric acid (max. 1%) (de Ge), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88067.AE 88067.AP
88067.AEEA 38.7 EUR
88067.AE 88067.AP
Germanium standard solution, 1000 mg/L Ge in 5% nitric acid with hydrofluoric acid (max. 1%) (de Ge), standard for ICP, Specpure®
Standaarden Kationische standaarden Germanium standaarden
Molecular Formula: Ge in 5% HNO₃/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085310
Certificaten: Traceable to NIST