Molybdenum standard solution, 1000 mg/L Mo in 5% nitric acid with hydrofluoric acid (max. 1%) (from Mo), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35758.AE
35758.AP
35758.AD
35758.AEEA
78
EUR
35758.AE
35758.AP
35758.AD
Molybdenum standard solution, 1000 mg/L Mo in 5% nitric acid with hydrofluoric acid (max. 1%) (from Mo), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Molybdeen standaarden
Molecular Formula: Mo in 5% HNO₃/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003465
Certificaten: Traceable to NIST