Molybdenum standard solution, 1000 mg/L Mo in 5% nitric acid with hydrofluoric acid (max. 1%) (from Mo), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35764.AE 35764.AP
35764.AEEA 43.7 EUR
35764.AE 35764.AP
Molybdenum standard solution, 1000 mg/L Mo in 5% nitric acid with hydrofluoric acid (max. 1%) (from Mo), standard for AAS, Specpure®
Standaarden Kationische standaarden Molybdeen standaarden
Molecular Formula: Mo in 5% HNO₃/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003465
Certificaten: Traceable to NIST
Order Now
Specificaties
Environmentally Preferable
MDL No. MFCD00003465