Niobium standard solution, 10000 mg/L Nb in 2% hydrofluoric acid (from NbCl₅), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14396.AD 14396.AE 14396.AP
14396.ADEA 91.1 EUR
14396.AD 14396.AE 14396.AP
Niobium standard solution, 10000 mg/L Nb in 2% hydrofluoric acid (from NbCl₅), standard for ICP, Specpure®
Standaarden Kationische standaarden Niobium standaarden
Molecular Formula: NbCl₅ in 2% HF
Physical Form: Liquid
UN No.: 1790
MDL No.: MFCD00011126
Certificaten: Traceable to NIST
Order Now
Specificaties
Environmentally Preferable
MDL No. MFCD00011126