Niobium standard solution, 1000 mg/L Nb in 2% hydrofluoric acid (from NbCl₅), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88083.AE
88083.AP
88083.AEEA
48
EUR
88083.AE
88083.AP
Niobium standard solution, 1000 mg/L Nb in 2% hydrofluoric acid (from NbCl₅), standard for AAS, Specpure®
Standaarden
Kationische standaarden
Niobium standaarden
Molecular Formula: NbCl₅ in 2% HF
Physical Form: Liquid
UN No.: 1790
MDL No.: MFCD00011126
Certificaten: Traceable to NIST