Palladium standard solution, 1000 mg/L Pd in 5% nitric acid (from Pd), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
44631.AD
44631.AE
44631.AP
44631.ADEA
105
EUR
44631.AD
44631.AE
44631.AP
Palladium standard solution, 1000 mg/L Pd in 5% nitric acid (from Pd), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Palladium standaarden
Molecular Formula: Pd in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011167
Certificaten: Traceable to NIST