Tungsten standard solution, 1000 mg/L W in 5% nitric acid with hydrofluoric acid (max. 1%) (from W), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
35770.AD
35770.AE
35770.AP
35770.ADEA
54.9
EUR
35770.AD
35770.AE
35770.AP
Tungsten standard solution, 1000 mg/L W in 5% nitric acid with hydrofluoric acid (max. 1%) (from W), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Wolfraam standaarden
Molecular Formula: W in 5% HNO₃/tr. HF
Physical Form: Liquid
Odour: Acidic
UN No.: 3264
MDL No.: MFCD00011461
Certificaten: Traceable to NIST