Hafnium standard solution, 1000 mg/L Hf in 5% hydrochloric acid (from HfOCl?), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13843.AD 13843.AE
13843.ADEA 62.6 EUR
13843.AD 13843.AE
Hafnium standard solution, 1000 mg/L Hf in 5% hydrochloric acid (from HfOCl?), standard for ICP, Specpure®
Standaarden Kationische standaarden Hafnium standaarden
Molecular Formula: HfClO₂ in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011032
Certificaten: Traceable to NIST