Phosphorus standard solution, 1,000 mg/L P in 5% nitric acid (from P), standard for AAS, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
89887.AE 89887.AP
89887.AEEA 39.5 EUR
89887.AE 89887.AP
Phosphorus standard solution, 1,000 mg/L P in 5% nitric acid (from P), standard for AAS, Specpure®
Standaarden Kationische standaarden Fosfor standaarden
Molecular Formula: P in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00133771
Certificaten: Traceable to NIST