Rhenium standard solution, 1000 mg/L Re in 5% nitric acid (from Re), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13817.AD
13817.AE
13817.AP
13817.ADEA
104
EUR
13817.AD
13817.AE
13817.AP
Rhenium standard solution, 1000 mg/L Re in 5% nitric acid (from Re), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Renium standaarden
Molecular Formula: Re in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011195
Certificaten: Traceable to NIST