Tantalum standard solution, 1000 mg/L Ta in 2% hydrofluoric acid (from TaCl₅), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13840.AD 13840.AE 13840.AP
13840.ADEA 70.3 EUR
13840.AD 13840.AE 13840.AP
Tantalum standard solution, 1000 mg/L Ta in 2% hydrofluoric acid (from TaCl₅), standard for ICP, Specpure®
Standaarden Kationische standaarden Tantalum standaarden
Molecular Formula: TaCl₅ in 2% HF
Physical Form: Liquid
UN No.: 1790
MDL No.: MFCD00011252
Certificaten: Traceable to NIST