2-Hydroxyhippuric acid 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: N-(2-Hydroxybenzoyl)glycine

158620025 158620100
ACRO158620025EA 79.3 EUR
ACRO158620025 ACRO158620100
2-Hydroxyhippuric acid 95%
2-Hydroxyhippuric acid
Formule: C₉H₉NO₄
Smeltpunt: 164…169 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002695
CAS nummer: 487-54-7
EINECS: 207-661-6

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 163 to 168 °C
HPLC ≥94 %
Calcium (Ca) ≤0.02 %
Iron (Fe) ≤0.005 %
Magnesium (Mg) ≤0.02 %
Potassium (K) ≤0.02 %
Heavy metals (as Pb) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR