4-Methoxyphenylhydrazinium chloride ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
4-Methoxyphenylhydrazine hydrochloride
L06076.06
L06076.14
L06076.22
L06076.06EA
47.7
EUR
L06076.06
L06076.14
L06076.22
4-Methoxyphenylhydrazinium chloride ≥99%
4-Methoxyphenylhydrazinium chloride
Purity: 99%
CAS No.: 19501-58-7
Molecular Formula: C7H10N2O.HCl
Formula Weight: 174.63
Melting Point: ca 161° dec.
Beilstein Reference: 3566583
Formule:
C₇H₁₁ClN₂O MW: 174.63 g/mol Smeltpunt: ∼ 161 °C (ontbinding) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00012945 CAS nummer: 19501-58-7 EINECS: 243-115-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...