Potassium heptafluoroniobate ≥99.5%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Potassium heptafluoroniobate(V)

51110.09 51110.18
51110.09EA 40.6 EUR
51110.09 51110.18
Potassium heptafluoroniobate ≥99.5%
Potassium heptafluoroniobate

Purity: 99.5% CAS No.: 16924-03-1 Molecular Formula: K2NbF7 Formula Weight: 304.10 Physical Form: Powder Sensitivity: Hygroscopic UN No.: 3288 MDL No.: MFCD00042368

Formule: F₇K₂Nb
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042368
CAS nummer: 16924-03-1
EINECS: 240-987-7
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR