Osmium standard solution, 1000 mg/L Os in 20% hydrochloric acid (from (NH₄)₂OsCl₆), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13871.AD
13871.AE
13871.AP
13871.ADEA
206
EUR
13871.AD
13871.AE
13871.AP
Osmium standard solution, 1000 mg/L Os in 20% hydrochloric acid (from (NH₄)₂OsCl₆), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Osmium standaarden
Molecular Formula: (NH₄)₂OsCl₆ in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011147
Certificaten: Traceable to NIST