Thulium standard solution, 1000 mg/L Tm in 5% nitric acid (from Tm₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13824.AD 13824.AE 13824.AP
13824.ADEA 87.5 EUR
13824.AD 13824.AE 13824.AP
Thulium standard solution, 1000 mg/L Tm in 5% nitric acid (from Tm₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Thulium standaarden
Molecular Formula: Tm₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011281
Certificaten: Traceable to NIST