Ethyl phenylsulphonylacetate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Ethyl 2-(phenylsulphonyl)acetate, Phenylsulphonylacetic acid Ethyl Ester
P1306-25G
P1306-5G
TCIAP1306-25GEA
183
EUR
TCIAP1306-25G
TCIAP1306-5G
Ethyl phenylsulphonylacetate ≥98.0% (by GC)
Ethyl phenylsulphonylacetate
Formule:
C₁₀H₁₂O₄S MW: 228.26 g/mol |
MDL Nummer:
MFCD00025041 CAS nummer: 7605-30-3 EINECS: 231-516-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 43.0 to 46.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...