Iron standard solution, concentrate, 1000 mg Fe in 15% hydrochloric acid (from FeCl₃) Titrisol® standard for AAS, Supelco®
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.09972.0001
1.09972.0050
1.09972.0001EA
34.9
EUR
1.09972.0001
1.09972.0050
Iron standard solution, concentrate, 1000 mg Fe in 15% hydrochloric acid (from FeCl₃) Titrisol® standard for AAS, Supelco®
Standaarden
Kationische standaarden
IJzer standaarden
Certificaten: Traceable to SRM from NIST