CryoTubes™, Nunc™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
373420 373530
479-2451EA 0 EUR
479-2451 479-2452
CryoTubes™, Nunc™
flesjes
Simplify sample inventory. Thermo Scientific™ Linear barcoded tubes feature Thermo Scientific™ Nunc™ and Thermo Scientific™ Nalgene™ Cryotubes with directly printed barcodes that are more robust than labels. Choose from available noncustom options or call for customised options in a wide range of choices of code type, length and format.
  • Black barcode directly printed onto white background
  • High contrast and readability even in high-frost conditions
  • Human readable code below the barcode for easy visual identification without a barcode reader
Barcodes resistant to IPA, DMSO, 5% bleach, 10% acetic acid and 10% NaOH, Noncytotoxic raw materials for added sample security and tubes certified free of DNase and RNase for genomic applications, External or internally threading options available, Certified SAL 10-6 for the highest sterility assurance available, Suitable for storage from general cold storage refrigeration (4 °C) down to the vapor phase of liquid nitrogen.