Disposable pestles for 1,5 ml microtubes, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F199230000 F199230001
BELAF199230000EA 255 EUR
BELAF199230000 BELAF199230001
Disposable pestles for 1,5 ml microtubes, SP Bel-Art
Vijzels en stampers Stampers
These disposable plastic (PP) pestles are excellent for re-suspending bacterial cells, precipitates of nucleic acids or proteins and pellets of other materials.
  • With 1,5 ml microcentrifuge tubes of the same shape for optimal grinding
  • Pestles also available without tubes