Stirrer blades, moon-shaped, with angular groove
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C401-02 C401-04 C401-08
BOHLC401-02EA 8.3 EUR
BOHLC401-02 BOHLC401-04 BOHLC401-08
Stirrer blades, moon-shaped, with angular groove
Roerders Roerbladen
Stirrer blades made of PTFE, with angular groove and 5,8 mm bore diameter.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Suitable for vessels with a 29/32 ground joint and glass stirrer shafts and KPG glass stirrer shafts.