Accessories for KL, KM, KS, SM and VKS shakers
Leverancier: Edmund Bühler GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0052059 0052058 0052056 0052057 0052201
444-9435EA 61.3 EUR
444-9435 444-9433 444-9431 444-9432 444-9434
Accessories for KL, KM, KS, SM and VKS shakers
Schudapparaten Schudapparaten met cirkelbeweging
The test tube racks can be fastened either on the standard rack system between the clamping strips or can be screwed on the universal tray by means of a hinged foot. Hinged foot is used to vary the angle of inclination of the test tubes. The microplate is tilted at approximately 20°