Racks for test tubes, indexed, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F187450023 F187450099
BELAF187450023EA 88.5 EUR
BELAF187450023 BELAF187450099
Racks for test tubes, indexed, SP Bel-Art
Rekken Tube Racks
White polypropylene, autoclavable
  • Excellent tool for clinical laboratories in identifying test tube locations for multiple samples
  • Typical applications include robotic sample processing equipment which uses 3 or 4 racks per unit
  • Available in either standard or serpentine numbering patterns
  • High visibility numbers are clearly silk-screened on the top of the rack
Racks designed for automated liquid handling systems, with round holes and tight tolerances hold 10 to 13 mm tubes, for accurate processing reducing lost samples and potential damage toMore Product Information